2-[4-(4-Methoxyphenyl)piperazin-1-yl]-2-methylpropanenitrile, tech

Lieferant: Apollo Scientific

APOSOR32825-1GEA 1390 EUR
APOSOR32825-1G
2-[4-(4-Methoxyphenyl)piperazin-1-yl]-2-methylpropanenitrile, tech
2-[4-(4-Methoxyphenyl)piperazin-1-yl]-2-methylpropanenitrile
Formel: C₁₅H₂₁N₃O
Molecular Weight: 259,347 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00202049
CAS-Nummer: 338754-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR