6-[(4-Methoxyphenyl)methyl]pyridazin-3-ol, tech
Lieferant: Apollo Scientific
APOSOR32818-1GEA
1390
EUR
APOSOR32818-1G
6-[(4-Methoxyphenyl)methyl]pyridazin-3-ol, tech
6-[(4-Methoxyphenyl)methyl]pyridazin-3-ol
Formel:
C₁₂H₁₂N₂O₂ Molecular Weight: 216,236 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00140698 CAS-Nummer: 338753-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...