(2E)-3-(Dimethylamino)-2-[4-(1H-pyrrol-1-yl)phenyl]prop-2-enenitrile, tech

Lieferant: Apollo Scientific

APOSOR32792-1GEA 1390 EUR
APOSOR32792-1G
(2E)-3-(Dimethylamino)-2-[4-(1H-pyrrol-1-yl)phenyl]prop-2-enenitrile, tech
(2E)-3-(Dimethylamino)-2-[4-(1H-pyrrol-1-yl)phenyl]prop-2-enenitrile
Formel: C₁₅H₁₅N₃
Molecular Weight: 237,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125523
CAS-Nummer: 866019-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR