3-Oxatricyclo[7.3.1.05,13]trideca-1(12),5(13),6,8,10-pentaen-2-one, tech

Lieferant: Apollo Scientific

APOSOR32710-1GEA 1390 EUR
APOSOR32710-1G
3-Oxatricyclo[7.3.1.05,13]trideca-1(12),5(13),6,8,10-pentaen-2-one, tech
3-Oxatricyclo[7.3.1.05,13]trideca-1(12),5(13),6,8,10-pentaen-2-one
Formel: C₁₂H₈O₂
Molecular Weight: 184,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138509
CAS-Nummer: 518-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR