2-(2,3-Dihydro-1,4-benzodioxin-6-yl)-hexahydro-1H-pyrrolo[1,2-c]imidazole-1,3-dione, tech
Lieferant: Apollo Scientific
APOSOR32693-1GEA
1390
EUR
APOSOR32693-1G
2-(2,3-Dihydro-1,4-benzodioxin-6-yl)-hexahydro-1H-pyrrolo[1,2-c]imidazole-1,3-dione, tech
2-(2,3-Dihydro-1,4-benzodioxin-6-yl)-hexahydro-1H-pyrrolo[1,2-c]imidazole-1,3-dione
Formel:
C₁₄H₁₄N₂O₄ Molecular Weight: 274,272 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26383446 CAS-Nummer: 1796902-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...