1-(Cyclopropylmethyl)-4-hydroxy-3-[(2E)-3-phenylprop-2-en-1-yl]-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

APOSOR32659-1GEA 1390 EUR
APOSOR32659-1G
1-(Cyclopropylmethyl)-4-hydroxy-3-[(2E)-3-phenylprop-2-en-1-yl]-1,2-dihydropyridin-2-one, tech
1-(Cyclopropylmethyl)-4-hydroxy-3-[(2E)-3-phenylprop-2-en-1-yl]-1,2-dihydropyridin-2-one
Formel: C₁₈H₁₉NO₂
Molecular Weight: 281,349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082727
CAS-Nummer: 478033-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR