4-Methoxy-2-(1H-pyrrol-1-yl)pyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

APOSOR32631-1GEA 1390 EUR
APOSOR32631-1G
4-Methoxy-2-(1H-pyrrol-1-yl)pyridine-3-carbonitrile, tech
4-Methoxy-2-(1H-pyrrol-1-yl)pyridine-3-carbonitrile
Formel: C₁₁H₉N₃O
Molecular Weight: 199,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975449
CAS-Nummer: 338412-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR