1,5,6-Trimethyl-1H-1,3-benzodiazol-4-amine, tech
Lieferant: Apollo Scientific
APOSOR32625-1GEA
1390
EUR
APOSOR32625-1G
1,5,6-Trimethyl-1H-1,3-benzodiazol-4-amine, tech
1,5,6-Trimethyl-1H-1,3-benzodiazol-4-amine
Formel:
C₁₀H₁₃N₃ Molecular Weight: 175,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00231368 CAS-Nummer: 338410-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...