2-[(Quinoxalin-2-yl)amino]ethan-1-ol, tech

Lieferant: Apollo Scientific

APOSOR32616-1GEA 1390 EUR
APOSOR32616-1G
2-[(Quinoxalin-2-yl)amino]ethan-1-ol, tech
2-[(Quinoxalin-2-yl)amino]ethan-1-ol
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243663
CAS-Nummer: 30466-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR