4-(2-Chlorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide, tech

Lieferant: Apollo Scientific

APOSOR32611-1GEA 1390 EUR
APOSOR32611-1G
4-(2-Chlorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide, tech
4-(2-Chlorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide
Formel: C₁₄H₁₃ClN₂O₂
Molecular Weight: 276,718 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141879
CAS-Nummer: 338404-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR