N,N-Dimethyl-4-(2-phenylacetyl)-1H-pyrrole-2-carboxamide, tech

Lieferant: Apollo Scientific

APOSOR32609-1GEA 1390 EUR
APOSOR32609-1G
N,N-Dimethyl-4-(2-phenylacetyl)-1H-pyrrole-2-carboxamide, tech
N,N-Dimethyl-4-(2-phenylacetyl)-1H-pyrrole-2-carboxamide
Formel: C₁₅H₁₆N₂O₂
Molecular Weight: 256,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141860
CAS-Nummer: 338403-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR