4-Oxo-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxaline-7-carbonitrile, tech
Lieferant: Apollo Scientific
APOSOR32588-1GEA
1390
EUR
APOSOR32588-1G
4-Oxo-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxaline-7-carbonitrile, tech
4-Oxo-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxaline-7-carbonitrile
Formel:
C₁₂H₁₁N₃O Molecular Weight: 213,235 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04124963 CAS-Nummer: 1008859-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...