2-tert-Butyl-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one, tech
Lieferant: Apollo Scientific
APOSOR32578-1GEA
1390
EUR
APOSOR32578-1G
2-tert-Butyl-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one, tech
2-tert-Butyl-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one
Formel:
C₁₀H₁₈N₂O₂ Molecular Weight: 198,262 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03848410 CAS-Nummer: 861210-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...