2-[4-(tert-Butoxy)phenyl]acetic acid, tech
Lieferant: Apollo Scientific
APOSOR32562-1GEA
1390
EUR
APOSOR32562-1G
2-[4-(tert-Butoxy)phenyl]acetic acid, tech
2-[4-(tert-Butoxy)phenyl]acetic acid
Formel:
C₁₂H₁₆O₃ Molecular Weight: 208,254 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411042 CAS-Nummer: 135066-21-6 |