Methyl (2-hydroxybenzoyl)imidoformate

Lieferant: Apollo Scientific

Gefahr

APOSOR3253-1GEA 109 EUR
APOSOR3253-1G
Methyl (2-hydroxybenzoyl)imidoformate
Methyl (2-hydroxybenzoyl)imidoformate
Formel: C₉H₉NO₃
Molecular Weight: 179,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998770
CAS-Nummer: 1000018-60-9
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR