2-[(3-Chloro-4-methoxyphenyl)methylidene]propanedinitrile, tech

Lieferant: Apollo Scientific

APOSOR32507-1GEA 1390 EUR
APOSOR32507-1G
2-[(3-Chloro-4-methoxyphenyl)methylidene]propanedinitrile, tech
2-[(3-Chloro-4-methoxyphenyl)methylidene]propanedinitrile
Formel: C₁₁H₇ClN₂O
Molecular Weight: 218,639 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03547576
CAS-Nummer: 362721-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR