4-[(6-Chloropyridin-3-yl)methoxy]benzonitrile, tech

Lieferant: Apollo Scientific

APOSOR32504-1GEA 1390 EUR
APOSOR32504-1G
4-[(6-Chloropyridin-3-yl)methoxy]benzonitrile, tech
4-[(6-Chloropyridin-3-yl)methoxy]benzonitrile
Formel: C₁₃H₉ClN₂O
Molecular Weight: 244,676 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124800
CAS-Nummer: 861207-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR