6H,7H,8H,9H,10H,12H-Azepino[2,1-b]quinazolin-12-one 95%
Lieferant: Apollo Scientific
APOSOR32495-1GEA
640
EUR
APOSOR32495-1G
6H,7H,8H,9H,10H,12H-Azepino[2,1-b]quinazolin-12-one 95%
6H,7H,8H,9H,10H,12H-Azepino[2,1-b]quinazolin-12-one
Formel:
C₁₃H₁₄N₂O Molecular Weight: 214,263 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00661748 CAS-Nummer: 4425-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...