9,10-Dimethoxy-1H,2H,3H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinoline-2,4-dione, tech

Lieferant: Apollo Scientific

APOSOR32478-1GEA 1390 EUR
APOSOR32478-1G
9,10-Dimethoxy-1H,2H,3H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinoline-2,4-dione, tech
9,10-Dimethoxy-1H,2H,3H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinoline-2,4-dione
Formel: C₁₅H₁₇NO₄
Molecular Weight: 275,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305515
CAS-Nummer: 5911-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR