3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester

Lieferant: Apollo Scientific

Achtung

APOSOR3246-1GEA 176 EUR
APOSOR3246-1G
3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester
3-Bromo-2-cyanobenzeneboronic acid neopentyl glycol ester
Formel: C₁₂H₁₃BBrNO₂
Molecular Weight: 293,952 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669549
CAS-Nummer: 883899-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR