(3E)-3-[2-(2-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one, tech
Lieferant: Apollo Scientific
APOSOR32419-1GEA
1390
EUR
APOSOR32419-1G
(3E)-3-[2-(2-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one, tech
(3E)-3-[2-(2-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one
Formel:
C₁₂H₁₅N₃O Molecular Weight: 217,267 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00794637 CAS-Nummer: 14490-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...