2-(2-Chlorobenzoyl)-1H-imidazole, tech
Lieferant: Apollo Scientific
APOSOR32408-1GEA
1390
EUR
APOSOR32408-1G
2-(2-Chlorobenzoyl)-1H-imidazole, tech
2-(2-Chlorobenzoyl)-1H-imidazole
Formel:
C₁₀H₇ClN₂O Molecular Weight: 206,628 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00125193 CAS-Nummer: 287196-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...