(3E)-1-(3,4-Dichlorophenyl)-3-(methoxyimino)propan-1-one, tech
Lieferant: Apollo Scientific
APOSOR32379-1GEA
1390
EUR
APOSOR32379-1G
(3E)-1-(3,4-Dichlorophenyl)-3-(methoxyimino)propan-1-one, tech
(3E)-1-(3,4-Dichlorophenyl)-3-(methoxyimino)propan-1-one
Formel:
C₁₀H₉Cl₂NO₂ Molecular Weight: 246,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00127284 CAS-Nummer: 320416-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...