4-[(5-Bromopyridin-3-yl)sulphonyl]morpholine 95%

Achtung

APOSOR3237-1GEA 0 EUR
APOSOR3237-1G
4-[(5-Bromopyridin-3-yl)sulphonyl]morpholine 95%
4-[(5-Bromopyridin-3-yl)sulphonyl]morpholine
Formel: C₉H₁₁BrN₂O₃S
Molecular Weight: 307,164 g/mol
Schmelzpunkt: 188…190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436023
CAS-Nummer: 889676-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR