1-(4-Acetylphenyl)-1H-pyrrole-2-carbaldehyde, tech

Lieferant: Apollo Scientific

APOSOR32342-1GEA 1390 EUR
APOSOR32342-1G
1-(4-Acetylphenyl)-1H-pyrrole-2-carbaldehyde, tech
1-(4-Acetylphenyl)-1H-pyrrole-2-carbaldehyde
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02661663
CAS-Nummer: 439094-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR