(4E)-4-[(Dimethylamino)methylidene]-2,3,4,5-tetrahydro-1-benzothiepin-5-one, tech

Lieferant: Apollo Scientific

APOSOR32332-1GEA 1390 EUR
APOSOR32332-1G
(4E)-4-[(Dimethylamino)methylidene]-2,3,4,5-tetrahydro-1-benzothiepin-5-one, tech
(4E)-4-[(Dimethylamino)methylidene]-2,3,4,5-tetrahydro-1-benzothiepin-5-one
Formel: C₁₃H₁₅NOS
Molecular Weight: 233,329 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082956
CAS-Nummer: 106364-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR