(Z)-(Ethyl N-[(Z)-benzoyl]ethanecarbohydrazonate), tech
Lieferant: Apollo Scientific
APOSOR32326-1GEA
1390
EUR
APOSOR32326-1G
(Z)-(Ethyl N-[(Z)-benzoyl]ethanecarbohydrazonate), tech
(Z)-(Ethyl N-[(Z)-benzoyl]ethanecarbohydrazonate)
Formel:
C₁₁H₁₄N₂O₂ Molecular Weight: 206,241 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02082701 CAS-Nummer: 85936-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...