3,5-Dihydroxypyridine 98%

APOSOR323256-1GEA 0 EUR
APOSOR323256-1G APOSOR323256-5G APOSOR323256-250MG
3,5-Dihydroxypyridine 98%
3,5-Dihydroxypyridine
Formel: C₅H₅NO₂
Molecular Weight: 111,099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01860225
CAS-Nummer: 3543-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR