4-Benzyl-2,6-dichloropyrimidine

Lieferant: Apollo Scientific

APOSOR323232-1GEA 1020 EUR
APOSOR323232-1G
4-Benzyl-2,6-dichloropyrimidine
4-Benzyl-2,6-dichloropyrimidine
Formel: C₁₁H₈Cl₂N₂
Molecular Weight: 239,101 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870615
CAS-Nummer: 796095-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR