(4-Bromothiophen-2-yl)-n,n-dimethylmethanamine

APOSOR323226-1GEA 0 EUR
APOSOR323226-1G
(4-Bromothiophen-2-yl)-n,n-dimethylmethanamine
(4-Bromothiophen-2-yl)-n,n-dimethylmethanamine
Formel: C₇H₁₀BrNS
Molecular Weight: 220,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16044590
CAS-Nummer: 78909-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR