2-(3,6-Dihydro-2,2-dimethyl-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Lieferant: Apollo Scientific

APOSOR323220-250MGEA 942 EUR
APOSOR323220-250MG
2-(3,6-Dihydro-2,2-dimethyl-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(3,6-Dihydro-2,2-dimethyl-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₃H₂₃BO₃
Molecular Weight: 238,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22683297
CAS-Nummer: 1215867-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR