4-(6H-Dibenzo[b,f]azocin-5-yl)-4-oxo-butyric acid methyl ester

Lieferant: Apollo Scientific

APOSOR323215-500MGEA 942 EUR
APOSOR323215-500MG
4-(6H-Dibenzo[b,f]azocin-5-yl)-4-oxo-butyric acid methyl ester
4-(6H-Dibenzo[b,f]azocin-5-yl)-4-oxo-butyric acid methyl ester
Formel: C₂₀H₁₉NO₃
Molecular Weight: 321,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28064327
CAS-Nummer: 1353016-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR