2-Chloro-2-(4'-methoxycarbonylphenylhydrazono)acetic acid ethyl ester

Lieferant: Apollo Scientific

APOSOR323214-1GEA 902 EUR
APOSOR323214-1G
2-Chloro-2-(4'-methoxycarbonylphenylhydrazono)acetic acid ethyl ester
2-Chloro-2-(4'-methoxycarbonylphenylhydrazono)acetic acid ethyl ester
Formel: C₁₂H₁₃ClN₂O₄
Molecular Weight: 284,696 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585466
CAS-Nummer: 1207062-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR