5-Cyclohexylpyrimidine-2,4,6(1h,3h,5h)-trione
Lieferant: Apollo Scientific
APOSOR323213-1GEA
902
EUR
APOSOR323213-1G
5-Cyclohexylpyrimidine-2,4,6(1h,3h,5h)-trione
5-Cyclohexylpyrimidine-2,4,6(1h,3h,5h)-trione
Formel:
C₁₀H₁₄N₂O₃ Molecular Weight: 210,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00576425 CAS-Nummer: 7391-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...