(Z)-Ethyl 2-(2-(5-(1-methyl-4-ethoxycarbonyl-1H-pyrazole))hydrazono)-2-chloroacetate

APOSOR323210-1GEA 0 EUR
APOSOR323210-1G
(Z)-Ethyl 2-(2-(5-(1-methyl-4-ethoxycarbonyl-1H-pyrazole))hydrazono)-2-chloroacetate
(Z)-Ethyl 2-(2-(5-(1-methyl-4-ethoxycarbonyl-1H-pyrazole))hydrazono)-2-chloroacetate
Formel: C₁₁H₁₅ClN₄O₄
Molecular Weight: 302,714 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585465
CAS-Nummer: 1207062-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR