1-O-Methyl 3-o-methyl 2-(tri-i-butylphosphoranylidene)propanedioate

Lieferant: Apollo Scientific

APOSOR323199-1GEA 807 EUR
APOSOR323199-1G
1-O-Methyl 3-o-methyl 2-(tri-i-butylphosphoranylidene)propanedioate
1-O-Methyl 3-o-methyl 2-(tri-i-butylphosphoranylidene)propanedioate
Formel: C₁₇H₃₃O₄P
Molecular Weight: 332,415 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18804137
CAS-Nummer: 501939-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR