4-Chloro-2-methoxypyridine-3-carbaldehyde

Lieferant: Apollo Scientific

APOSOR323182-250MGEA 234 EUR
APOSOR323182-250MG APOSOR323182-1G APOSOR323182-100MG
4-Chloro-2-methoxypyridine-3-carbaldehyde
4-Chloro-2-methoxypyridine-3-carbaldehyde
Formel: C₇H₆ClNO₂
Molecular Weight: 171,581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13188720
CAS-Nummer: 1008451-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR