4-Chloro-2-methoxypyridine-3-carbonitrile 98%

Lieferant: Apollo Scientific

Gefahr

APOSOR323180-1GEA 669 EUR
APOSOR323180-1G APOSOR323180-250MG
4-Chloro-2-methoxypyridine-3-carbonitrile 98%
4-Chloro-2-methoxypyridine-3-carbonitrile
Formel: C₇H₅ClN₂O
Molecular Weight: 168,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585278
CAS-Nummer: 1008451-56-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR