Diethyl 6-bromohexylphosphonate

Lieferant: Apollo Scientific

APOSOR323174-500MGEA 757 EUR
APOSOR323174-500MG
Diethyl 6-bromohexylphosphonate
Diethyl 6-bromohexylphosphonate
Formel: C₁₀H₂₂BrO₃P
Molecular Weight: 301,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00135788
CAS-Nummer: 100462-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR