3-(Cyclopentanecarboxamido)-[1,2,4]triazolo[4,3-a]pyridine-8-carboxylic acid

Lieferant: Apollo Scientific

APOSOR323171-500MGEA 734 EUR
APOSOR323171-500MG
3-(Cyclopentanecarboxamido)-[1,2,4]triazolo[4,3-a]pyridine-8-carboxylic acid
3-(Cyclopentanecarboxamido)-[1,2,4]triazolo[4,3-a]pyridine-8-carboxylic acid
Formel: C₁₃H₁₄N₄O₃
Molecular Weight: 274,275 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585339
CAS-Nummer: 1206970-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR