4-(Morpholinomethyl)phenol 97%

Lieferant: Apollo Scientific

APOSOR323166-500MGEA 452 EUR
APOSOR323166-500MG APOSOR323166-1G
4-(Morpholinomethyl)phenol 97%
4-(Morpholinomethyl)phenol
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01467510
CAS-Nummer: 80166-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR