3-((Propylsulphonyl)methyl)piperidine

Lieferant: Apollo Scientific

APOSOR323165-250MGEA 734 EUR
APOSOR323165-250MG
3-((Propylsulphonyl)methyl)piperidine
3-((Propylsulphonyl)methyl)piperidine
Formel: C₉H₁₉NO₂S
Molecular Weight: 205,318 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10017386
CAS-Nummer: 1018533-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR