Methyl 3-(2-(pyridin-4-yl)acetamido)-2-aminobenzoate

Lieferant: Apollo Scientific

APOSOR323164-250MGEA 734 EUR
APOSOR323164-250MG
Methyl 3-(2-(pyridin-4-yl)acetamido)-2-aminobenzoate
Methyl 3-(2-(pyridin-4-yl)acetamido)-2-aminobenzoate
Formel: C₁₅H₁₅N₃O₃
Molecular Weight: 285,298 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23378556
CAS-Nummer: 1421933-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR