2-Chloro-2-(4'-cyanophenylhydrazono)acetic acid ethyl ester

APOSOR323162-1GEA 0 EUR
APOSOR323162-1G
2-Chloro-2-(4'-cyanophenylhydrazono)acetic acid ethyl ester
2-Chloro-2-(4'-cyanophenylhydrazono)acetic acid ethyl ester
Formel: C₁₁H₁₀ClN₃O₂
Molecular Weight: 251,669 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585464
CAS-Nummer: 1595286-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR