(S)-2-Acetamido-6-(diphenyl(p-tolyl)methylamino)hexanoic acid

Lieferant: Apollo Scientific

APOSOR323155-500MGEA 713 EUR
APOSOR323155-500MG
(S)-2-Acetamido-6-(diphenyl(p-tolyl)methylamino)hexanoic acid
(S)-2-Acetamido-6-(diphenyl(p-tolyl)methylamino)hexanoic acid
Formel: C₂₈H₃₂N₂O₃
Molecular Weight: 444,565 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18157736
CAS-Nummer: 1565306-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR