3,3-Di-(phthalimidomethyl)oxetane
Lieferant: Apollo Scientific
APOSOR323151-1GEA
713
EUR
APOSOR323151-1G
3,3-Di-(phthalimidomethyl)oxetane
3,3-Di-(phthalimidomethyl)oxetane
Formel:
C₂₁H₁₆N₂O₅ Molecular Weight: 376,362 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14585336 CAS-Nummer: 198572-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...