1,3,7-Triaza-spiro[4.5]decane-2,4-dione

Lieferant: Apollo Scientific

APOSOR323145-500MGEA 671 EUR
APOSOR323145-500MG
1,3,7-Triaza-spiro[4.5]decane-2,4-dione
1,3,7-Triaza-spiro[4.5]decane-2,4-dione
Formel: C₇H₁₁N₃O₂
Molecular Weight: 169,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07776799
CAS-Nummer: 78222-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR