Methyl 5-bromo-7-methoxybenzofuran-2-carboxylate

Lieferant: Apollo Scientific

APOSOR323134-500MGEA 671 EUR
APOSOR323134-500MG
Methyl 5-bromo-7-methoxybenzofuran-2-carboxylate
Methyl 5-bromo-7-methoxybenzofuran-2-carboxylate
Formel: C₁₁H₉BrO₄
Molecular Weight: 285,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987551
CAS-Nummer: 1242240-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR