tert-Butyl 6-(2,5-dioxo-2H-pyrrol-1(5H)-yl)hexylcarbamate

Lieferant: Apollo Scientific

APOSOR323132-250MGEA 671 EUR
APOSOR323132-250MG
tert-Butyl 6-(2,5-dioxo-2H-pyrrol-1(5H)-yl)hexylcarbamate
tert-Butyl 6-(2,5-dioxo-2H-pyrrol-1(5H)-yl)hexylcarbamate
Formel: C₁₅H₂₄N₂O₄
Molecular Weight: 296,362 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22683316
CAS-Nummer: 124529-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR