5-Amino-2-methoxyphenyl pivalate

Lieferant: Apollo Scientific

APOSOR323130-500MGEA 669 EUR
APOSOR323130-500MG
5-Amino-2-methoxyphenyl pivalate
5-Amino-2-methoxyphenyl pivalate
Formel: C₁₂H₁₇NO₃
Molecular Weight: 223,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27635169
CAS-Nummer: 660870-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR